Furfuryl acetate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Furan-2-ylmethyl acetate, Acetic acid furfuryl ester
M-Clarity™ quality level = MQ300
W249009-10KG-K
W249009-1KG-K
W249009-5KG-K
SIALW249009-10KG-KEA
25610
CZK
SIALW249009-10KG-K
SIALW249009-1KG-K
SIALW249009-5KG-K
Furfuryl acetate, Sigma-Aldrich®
Furfuryl acetate
Vzorec:
C₇H₈O₃ MW: 140.14 Bod varu: 175 Hustota: 1.1176 g/cm³ (20 °C) |
MDL Number:
MFCD00003251 CAS číslo: 623-17-6 EINECS: 210-775-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...