Tisk…

Ethyl isovalerate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Ethyl isopentanoate, Isovaleric acid ethyl ester, 3-Methylbutyric acid ethyl ester, Isopropyl acetic acid ethyl ester, 3-Methylbutanoic acid ethyl ester

M-Clarity™ quality level = MQ300

W246301-1KG-K W246301-20KG-K W246301-4KG-K
SIALW246301-1KG-KEA 2050 CZK
SIALW246301-1KG-K SIALW246301-20KG-K SIALW246301-4KG-K
Ethyl isovalerate, Sigma-Aldrich®
Ethyl isovalerate
Vzorec: C₇H₁₄O₂
MW: 130,19 g/mol
Bod varu: 131…133 °C (1013 hPa)
Bod tání: –99 °C
Hustota: 0,86 g/cm³ (20 °C)
MDL Number: MFCD00009203
CAS číslo: 108-64-5
EINECS: 203-602-3
UN: 3272
ADR: 3,III
Merck Index: 13,03849

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR