Benzyl mercaptan, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Toluene-α-thiol, α-Toluenethiol
M-Clarity™ quality level = MQ300
W214701-10KG-K
W214701-1KG-K
SIALW214701-10KG-KEA
59490
CZK
SIALW214701-10KG-K
SIALW214701-1KG-K
Benzyl mercaptan, Sigma-Aldrich®
Benzyl mercaptan
Vzorec:
C₇H₈S MW: 124,21 g/mol Bod varu: 195 °C (1013 hPa) Bod tání: 29 °C Hustota: 1,055 g/cm³ (20 °C) Bod vzplanutí: 70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004867 CAS číslo: 100-53-8 EINECS: 202-862-5 UN: 2810 ADR: 6.1,II Merck Index: 12,09456 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...