Tisk…

3,4-Dimethoxybenzylamine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Veratrylamine

M-Clarity™ quality level = MQ100

V1309-25G V1309-5G
SIALV1309-25GEA 4850 CZK
SIALV1309-25G SIALV1309-5G
3,4-Dimethoxybenzylamine, Sigma-Aldrich®
3,4-Dimethoxybenzylamine
Vzorec: C₉H₁₃NO₂
MW: 167,21 g/mol
Bod varu: 154…158 °C (12 mmHg)
Hustota: 1,109 g/cm³ (20 °C)
MDL Number: MFCD00008116
CAS číslo: 5763-61-1
EINECS: 227-287-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR