Tisk…

Trityl bromide, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Bromotriphenylmethane, Triphenylbromomethane, Triphenylmethyl bromide

M-Clarity™ quality level = MQ100

T83607-100G T83607-25G
SIALT83607-100GEA 7240 CZK
SIALT83607-100G SIALT83607-25G
Trityl bromide, Sigma-Aldrich®
Trityl bromide
Vzorec: (C₆H₅)₃CBr
Bod varu: 230 °C (1013 hPa)
Hustota: 1,55 g/cm³ (20 °C)
MDL Number: MFCD00000120
CAS číslo: 596-43-0
EINECS: 209-884-4
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR