Tisk…

Trimethylsulphoxonium iodide, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Trimethyloxosulphonium iodide

M-Clarity™ quality level = MQ100

T80500-100G T80500-25G T80500-500G
SIALT80500-100GEA 2390 CZK
SIALT80500-100G SIALT80500-25G SIALT80500-500G
Trimethylsulphoxonium iodide, Sigma-Aldrich®
Trimethylsulphoxonium iodide
Vzorec: C₃H₉IOS
MW: 220,07 g/mol
Bod tání: 165…175 °C
MDL Number: MFCD00011899
CAS číslo: 1774-47-6
EINECS: 217-204-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR