Tricin, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)acetic acid, Tricine buffer, N-(Tris(hydroxymethyl)methyl)glycine
M-Clarity™ quality level = MQ400
T5816-100G
T5816-1KG
T5816-25G
T5816-500G
SIALT5816-100GEA
6320
CZK
SIALT5816-100G
SIALT5816-1KG
SIALT5816-25G
SIALT5816-500G
Tricin, Sigma-Aldrich®
Tricine
Vzorec:
(HOCH₂)₃CNHCH₂CO₂H MW: 179,17 g/mol Bod tání: 187 °C Hustota: 1,523 g/cm³ (19.85 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004277 CAS číslo: 5704-04-1 EINECS: 227-193-6 Merck Index: 13,09724 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...