Tisk…

p-Tolylacetic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-(p-Tolyl)acetic acid, 4-Tolylacetic acid, (4-Methylphenyl)acetic acid

M-Clarity™ quality level = MQ100

T38105-25G T38105-5G
SIALT38105-25GEA 2430 CZK
SIALT38105-25G SIALT38105-5G
p-Tolylacetic acid, Sigma-Aldrich®
p-Tolylacetic acid
Vzorec: (C₆H₄(CH₃))CH₂COOH
MW: 150,18 g/mol
Bod varu: 265 °C (1013 hPa)
Bod tání: 91 °C
MDL Number: MFCD00004353
CAS číslo: 622-47-9
EINECS: 210-738-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR