Tisk…

Thymine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 5-Methylpyrimidine-2,4(1H,3H)-dione, 5-Methyluracil

M-Clarity™ quality level = MQ200

T0895-25G T0895-5G
SIALT0895-25GEA 5110 CZK
SIALT0895-25G SIALT0895-5G
Thymine, Sigma-Aldrich®
Thymine
Vzorec: C₅H₆N₂O₂
MW: 126,11 g/mol
Bod varu: 389 °C (1013 hPa)
Bod tání: 218 °C
Hustota: 1,454 g/cm³ (25 °C)
MDL Number: MFCD00006026
CAS číslo: 65-71-4
EINECS: 200-616-1
Merck Index: 12,09539

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR