Tisk…
Tricin, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: 2-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)acetic acid, Tricine buffer, N-(Tris(hydroxymethyl)methyl)glycine
M-Clarity™ quality level = MQ400
T0377-100G T0377-1KG T0377-250G T0377-25G T0377-25KG T0377-5KG
SIALT0377-100GEA 6480 CZK
SIALT0377-100G SIALT0377-1KG SIALT0377-250G SIALT0377-25G SIALT0377-25KG SIALT0377-5KG
Tricin, Sigma-Aldrich®
Tricin
Vzorec: (HOCH₂)₃CNHCH₂CO₂H
MW: 179.17
Bod tání: 187
Hustota: 1.523 g/cm³ (19.85 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004277
CAS číslo: 5704-04-1
EINECS: 227-193-6
Merck Index: 13,09724