D(-)-Ribóza, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
R1757-10G-A
R1757-100G-A
R1757-25G-A
SIALR1757-10G-AEA
780
CZK
SIALR1757-10G-A
SIALR1757-100G-A
SIALR1757-25G-A
D(-)-Ribóza, Sigma-Aldrich®
D(-)-Ribose
Vzorec:
C₅H₁₀O₅ MW: 150,13 g/mol Bod varu: 331 °C (1013 hPa) Bod tání: 90…95 °C Hustota: 0,8 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00135453 CAS číslo: 50-69-1 EINECS: 200-059-4 Merck Index: 12,08371 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...