5-Methylphenazinium methyl sulphate, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ200
P9625-500MG
P9625-5G
P9625-10G
P9625-1G
P9625-25G
SIALP9625-500MGEA
1790
CZK
SIALP9625-500MG
SIALP9625-5G
SIALP9625-10G
SIALP9625-1G
SIALP9625-25G
5-Methylphenazinium methyl sulphate, Sigma-Aldrich®
5-Methylphenazinium methyl sulphate
Vzorec:
C₁₄H₁₄N₂O₄S Bod tání: 198 °C Storage Temperature: Freezer |
MDL Number:
MFCD00011923 CAS číslo: 299-11-6 EINECS: 206-072-1 Merck Index: 12,06185 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...