Propiolic acid, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Acetylenecarboxylic Acid, 2-Propynoic acid
M-Clarity™ quality level = MQ100
P51400-100G
P51400-25G
P51400-5G
SIALP51400-100GEA
9000
CZK
SIALP51400-100G
SIALP51400-25G
SIALP51400-5G
Propiolic acid, Sigma-Aldrich®
Propiolic acid
Vzorec:
C₃H₂O₂ MW: 70,05 g/mol Bod varu: 140 °C (1013 hPa) Bod tání: 18 °C Hustota: 1,136 g/cm³ (20 °C) Bod vzplanutí: 58 °C |
MDL Number:
MFCD00004360 CAS číslo: 471-25-0 EINECS: 207-437-8 UN: 2920 ADR: 8,II Merck Index: 12,08006 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...