Tisk…

Phenylpropiolic acid, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: Phenylacetylenecarboxylic acid, Phenylpropynoic acid, 3-Phenylpropiolic acid

M-Clarity™ quality level = MQ100

P31205-25G P31205-5G
SIALP31205-25GEA 5530 CZK
SIALP31205-25G SIALP31205-5G
Phenylpropiolic acid, Sigma-Aldrich®
Phenylpropiolic acid
Vzorec: C₉H₆O₂
MW: 146,15 g/mol
Bod varu: 294 °C (1013 hPa)
Bod tání: 135…138 °C
Hustota: 1,28 g/cm³ (20 °C)
MDL Number: MFCD00004361
CAS číslo: 637-44-5
EINECS: 211-285-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR