Tisk…

N-Phenylmaleimide, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

P27100-100G P27100-25G
SIALP27100-100GEA 5220 CZK
SIALP27100-100G SIALP27100-25G
N-Phenylmaleimide, Sigma-Aldrich®
N-Phenylmaleimide
Vzorec: C₁₀H₇NO₂
MW: 173,17 g/mol
Bod varu: 299 °C (1013 hPa)
Bod tání: 84…86 °C
MDL Number: MFCD00005502
CAS číslo: 941-69-5
EINECS: 213-382-0
UN: 2811
ADR: 6.1,III
Merck Index: 12,07452

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR