Tisk…

Oleyl alcohol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: cis-Octadec-9-en-1-ol, cis-9-Octadecen-1-ol

M-Clarity™ quality level = MQ200

O8880-1G O8880-5G
SIALO8880-1GEA 8200 CZK
SIALO8880-1G SIALO8880-5G
Oleyl alcohol, Sigma-Aldrich®
Oleyl alcohol
Vzorec: C₁₈H₃₆O
MW: 268,48 g/mol
Bod varu: 333 °C (1013 hPa)
Bod tání: 13…19 °C
Hustota: 0,85 g/cm³ (20 °C)
MDL Number: MFCD00002993
CAS číslo: 143-28-2
EINECS: 205-597-3
Merck Index: 12,06968

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR