6-Methyl-5-hepten-2-one, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-Methyl-2-hepten-6-one, Sulcatone
M-Clarity™ quality level = MQ100
M48805-100ML
M48805-5ML
SIALM48805-100MLEA
1020
CZK
SIALM48805-100ML
SIALM48805-5ML
6-Methyl-5-hepten-2-one, Sigma-Aldrich®
6-Methyl-5-hepten-2-one
Vzorec:
C₈H₁₄O Bod varu: 173 °C (1013 hPa) Hustota: 0,85 g/cm³ (20 °C) Bod vzplanutí: 57 °C |
MDL Number:
MFCD00008905 CAS číslo: 110-93-0 EINECS: 203-816-7 UN: 1224 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...