4-Methylpyrocatechol, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
M34200-100G
M34200-500G
M34200-5G
SIALM34200-100GEA
3200
CZK
SIALM34200-100G
SIALM34200-500G
SIALM34200-5G
4-Methylpyrocatechol, Sigma-Aldrich®
4-Methylpyrocatechol
Vzorec:
C₇H₈O₂ |
MDL Number:
MFCD00002205 CAS číslo: 452-86-8 EINECS: 207-214-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...