(3-Methoxyphenyl)acetic acid, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
3-Methoxyphenylacetic acid
M-Clarity™ quality level = MQ100
M19007-25G
M19007-5G
SIALM19007-25GEA
3530
CZK
SIALM19007-25G
SIALM19007-5G
(3-Methoxyphenyl)acetic acid, Sigma-Aldrich®
(3-Methoxyphenyl)acetic acid
Vzorec:
(C₆H₄(OCH₃))CH₂COOH Bod varu: 305,95 °C (1013 hPa) Bod tání: 71…73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004334 CAS číslo: 1798-09-0 EINECS: 217-282-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...