Lithocholic acid, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
L6250-10G
L6250-25G
SIALL6250-10GEA
2890
CZK
SIALL6250-10G
SIALL6250-25G
Lithocholic acid, Sigma-Aldrich®
Lithocholic acid
Vzorec:
C₂₄H₄₀O₃ Bod tání: 190 °C |
MDL Number:
MFCD00003682 CAS číslo: 434-13-9 EINECS: 207-099-1 Merck Index: 12,05571 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...