Tisk…

Kyselina 5-hydroxyindol-3-yl-octová, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

H8876-100MG H8876-1G H8876-500MG
SIALH8876-100MGEA 2180 CZK
SIALH8876-100MG SIALH8876-1G SIALH8876-500MG
Kyselina 5-hydroxyindol-3-yl-octová, Sigma-Aldrich®
Kyselina 5-hydroxyindol-3-yl-octová
Vzorec: C₁₀H₉NO₃
Bod tání: 166 °C
Storage Temperature: Freezer
MDL Number: MFCD00005639
CAS číslo: 54-16-0
EINECS: 200-195-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR