N-Hydroxyphthalimide, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-Hydroxyisoindoline-1,3-dione, 2-Hydroxy-1H-isoindole-1,3(2H)-dione, N-Hydroxypthalimide
M-Clarity™ quality level = MQ200
H53704-100G
H53704-500G
H53704-5G
SIALH53704-100GEA
1170
CZK
SIALH53704-100G
SIALH53704-500G
SIALH53704-5G
N-Hydroxyphthalimide, Sigma-Aldrich®
N-Hydroxyphthalimide
Vzorec:
C₈H₅NO₃ Bod varu: 412 °C (1013 hPa) Bod tání: 230…233 °C Hustota: 1,58 g/cm³ (20 °C) |
MDL Number:
MFCD00005891 CAS číslo: 524-38-9 EINECS: 208-358-1 Merck Index: 13,04864 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...