Tisk…

Ethylurea, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

E51007-100G E51007-25G
SIALE51007-100GEA 4590 CZK
SIALE51007-100G SIALE51007-25G
Ethylurea, Sigma-Aldrich®
Ethylurea
Vzorec: C₃H₈N₂O
Bod varu: 258 °C (1013 hPa)
Hustota: 1,213 g/cm³ (18 °C)
MDL Number: MFCD00007953
CAS číslo: 625-52-5
EINECS: 210-898-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR