Tisk…

N-ethylmaleimid (NEM), Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: N-Ethylmaleimide, 1-Ethyl-1H-pyrrole-2,5-dione

M-Clarity™ quality level = MQ200

E3876-100G E3876-25G E3876-5G
SIALE3876-100GEA 39040 CZK
SIALE3876-100G SIALE3876-25G SIALE3876-5G
N-ethylmaleimid (NEM), Sigma-Aldrich®
N-ethylmaleimid (NEM)
Vzorec: C₆H₇NO₂
Bod varu: 210 °C (1013 hPa)
Bod tání: 46 °C
MDL Number: MFCD00005509
CAS číslo: 128-53-0
EINECS: 204-892-4
UN: 2928
ADR: 6.1,II
Merck Index: 13,03855

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR