EDTA tripotassium salt dihydrate, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
Ethylenedinitrilotetra-acetic acid tripotassium salt dihydrate, Tripotassium hydrogen ethylenediaminetetraacetate dihydrate, Ethylenediaminetetra-acetic acid tripotassium salt dihydrate, Potassium 2,2'-((2-((carboxylatomethyl)(carboxymethyl)amino)ethyl)azanediyl)diacetate dihydrate, Edetic acid tripotassium salt dihydrate
MQ200
E0270-100G
E0270-25G
SIALE0270-100GEA
11610
CZK
SIALE0270-100G
SIALE0270-25G
EDTA tripotassium salt dihydrate, Sigma-Aldrich®
EDTA tripotassium salt dihydrate
Vzorec:
C₁₀H₁₃K₃N₂O₈·2H₂O MW: 442.55 Storage Temperature: Ambient |
MDL Number:
MFCD00149672 CAS číslo: 65501-24-8 EINECS: 241-543-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...