Tisk…

2-Deoxy-D(-)-ribóza, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Deoxy-D(-)-erythropentose, Thyminose, 2-Deoxy-D(-)-arabinose, 2'-Deoxy-D-Ribose, (3S,4R)-3,4,5-Trihydroxypentanal

M-Clarity™ quality level = MQ200

D5899-10MG D5899-1G
SIALD5899-10MGEA 859 CZK
SIALD5899-10MG SIALD5899-1G
2-Deoxy-D(-)-ribóza, Sigma-Aldrich®
2-Deoxy-D(-)-ribose
Vzorec: C₅H₁₀O₄
MW: 134,13 g/mol
Bod varu: 292 °C (1013 hPa)
Bod tání: 95…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00135904
CAS číslo: 533-67-5
EINECS: 208-573-0
Merck Index: 13,02925

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR