Tisk…

Deoxycholic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 12α-Hydroxylithocholic acid, (R)-4-((3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-Dihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoic acid, 3α,12α-Dihydroxy-5β-cholanic acid, 3α,12α-Dihydroxy-5β-cholan-24-oic acid, DCA, 12alpha-Hydroxylithocholic acid, 3α,12α-Dihydroxycholan-24-oic acid, 3α,12α-Dihydroxycholanic acid

M-Clarity™ quality level = MQ500

D2510-100G D2510-10G D2510-500G
SIALD2510-100GEA 5680 CZK
SIALD2510-100G SIALD2510-10G SIALD2510-500G
Deoxycholic acid, Sigma-Aldrich®
Deoxycholic acid
Vzorec: C₂₄H₄₀O₄
Bod varu: 504 °C (1013 hPa)
Bod tání: 190 °C
MDL Number: MFCD00003673
CAS číslo: 83-44-3
EINECS: 201-478-5
Merck Index: 13,02916

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR