Tisk…

1,3-Dimethyl-2-thiourea, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1,3-Dimethylthiourea, N,N'-Dimethylthiourea

M-Clarity™ quality level = MQ200

D188700-100G D188700-5G
SIALD188700-100GEA 1080 CZK
SIALD188700-100G SIALD188700-5G
1,3-Dimethyl-2-thiourea, Sigma-Aldrich®
1,3-Dimethyl-2-thiourea
Vzorec: C₃H₈N₂S
MW: 104,18 g/mol
Bod varu: 133 °C (1013 hPa)
MDL Number: MFCD00004923
CAS číslo: 534-13-4
EINECS: 208-588-2
Merck Index: 13,03287

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR