1,4-Dimethylpiperazine, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
sym-Dimethylpiperazine
M-Clarity™ quality level = MQ100
D179302-100G
D179302-5G
SIALD179302-100GEA
1740
CZK
SIALD179302-100G
SIALD179302-5G
1,4-Dimethylpiperazine, Sigma-Aldrich®
1,4-Dimethylpiperazine
Vzorec:
C₆H₁₄N₂ MW: 114,19 g/mol Bod varu: 131 °C (1013 hPa) Bod tání: –1 °C Hustota: 0,85 g/cm³ (20 °C) |
MDL Number:
MFCD00006156 CAS číslo: 106-58-1 EINECS: 203-412-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...