Chenodeoxycholic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
3α,7α-Dihydroxycholanic acid, 7α-Hydroxylithocholic acid, 7alpha-Hydroxylithocholic acid, 3α,7α-Dihydroxy-5β-cholan-24-oic acid, 3α,7α-Dihydroxycholan-24-oic acid, Chenodiol, 3α,7α-Dihydroxy-5β-cholanic acid
M-Clarity™ quality level = MQ200
C9377-100MG
C9377-25G
C9377-5G
SIALC9377-100MGEA
1440
CZK
SIALC9377-100MG
SIALC9377-25G
SIALC9377-5G
Chenodeoxycholic acid, Sigma-Aldrich®
Chenodeoxycholic acid
Vzorec:
C₂₄H₄₀O₄ Hustota: 1,162 g/cm³ (20 °C) |
MDL Number:
MFCD00064142 CAS číslo: 474-25-9 EINECS: 207-481-8 Merck Index: 12,02096 |