Tisk…
Sodium cholate hydrate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: 3α,7α,12α-Trihydroxycholanic acid sodium salt hydrate, Sodium 3α,7α,12α-trihydroxy-5β-cholanoate hydrate, 3α,7α,12α-Trihydroxycholan-24-oic acid sodium salt hydrate, Cholic acid sodium salt hydrate, Sodium 3alpha,7alpha,12alpha-trihydroxy-5beta-cholanoate hydrate, 3α,7α,12α-Trihydroxy-5β-cholanic acid sodium salt hydrate, 3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid sodium salt hydrate
M-Clarity™ quality level = MQ200
C9282-100G C9282-25G
SIALC9282-100GEA 7470 CZK
SIALC9282-100G SIALC9282-25G
Sodium cholate hydrate, Sigma-Aldrich®
Sodium cholate hydrate
Vzorec: C₂₄H₃₉NaO₅·H₂O
MW: 448,58 g/mol
MDL Number: MFCD00064138
CAS číslo: 206986-87-0
EINECS: 206-643-5