Eukalyptol, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
1,8-Cineole, 1,3,3-Trimethyl-2-oxabicyclo[2.2.2]octane
M-Clarity™ quality level = MQ200
C80601-100ML
C80601-500ML
C80601-5ML
SIALC80601-100MLEA
1150
CZK
SIALC80601-100ML
SIALC80601-500ML
SIALC80601-5ML
Eukalyptol, Sigma-Aldrich®
Eukalyptol
Vzorec:
C₁₀H₁₈O Bod varu: 176…177 °C (1013 hPa) Hustota: 0,92 g/cm³ (20 °C) Bod vzplanutí: 46 °C Storage Temperature: Ambient |
MDL Number:
MFCD00167977 CAS číslo: 470-82-6 EINECS: 207-431-5 UN: 3271 ADR: 3,III Merck Index: 12,03940 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...