Tisk…

Sodium cholate hydrate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3α,7α,12α-Trihydroxycholanic acid sodium salt hydrate, Sodium 3α,7α,12α-trihydroxy-5β-cholanoate hydrate, 3α,7α,12α-Trihydroxycholan-24-oic acid sodium salt hydrate, Cholic acid sodium salt hydrate, Sodium 3alpha,7alpha,12alpha-trihydroxy-5beta-cholanoate hydrate, 3α,7α,12α-Trihydroxy-5β-cholanic acid sodium salt hydrate, 3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid sodium salt hydrate

M-Clarity™ quality level = MQ200

C6445-100G C6445-10G C6445-25G C6445-500G
SIALC6445-100GEA 18520 CZK
SIALC6445-100G SIALC6445-10G SIALC6445-25G SIALC6445-500G
Sodium cholate hydrate, Sigma-Aldrich®
Sodium cholate hydrate
Vzorec: C₂₄H₃₉NaO₅·H₂O
MW: 448,58 g/mol
MDL Number: MFCD00064138
CAS číslo: 206986-87-0
EINECS: 206-643-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR