4-Chloro-4'-fluorobutyrophenone, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
4-Chloro-1-(4-fluorophenyl)butan-1-one, ω-Chloro-4-fluorobutyrophenone, γ-Chloro-4-fluorobutyrophenone
M-Clarity™ quality level = MQ100
C43609-100G
C43609-25G
SIALC43609-100GEA
2870
CZK
SIALC43609-100G
SIALC43609-25G
4-Chloro-4'-fluorobutyrophenone, Sigma-Aldrich®
4-Chloro-4'-fluorobutyrophenone
Vzorec:
C₁₀H₁₀ClFO MW: 200,64 g/mol Bod varu: 300 °C (1013 hPa) Hustota: 1,2267 g/cm³ (20 °C) |
MDL Number:
MFCD00001007 CAS číslo: 3874-54-2 EINECS: 223-395-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...