Benzopinacol, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
B9807-100G
B9807-10G
SIALB9807-100GEA
4220
CZK
SIALB9807-100G
SIALB9807-10G
Benzopinacol, Sigma-Aldrich®
Benzopinacol
Vzorec:
(C₆H₅)₂C(OH)C(OH)(C₆H₅)₂ Bod varu: 508 °C (1013 hPa) Bod tání: 170…175 °C Hustota: 1,24 g/cm³ (20 °C) |
MDL Number:
MFCD00004448 CAS číslo: 464-72-2 EINECS: 207-356-8 Merck Index: 13,01101 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...