N-(3-Bromopropyl)phthalimide, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
B80003-100G
B80003-25G
SIALB80003-100GEA
3610
CZK
SIALB80003-100G
SIALB80003-25G
N-(3-Bromopropyl)phthalimide, Sigma-Aldrich®
N-(3-Bromopropyl)phthalimide
Vzorec:
C₁₁H₁₀BrNO₂ MW: 268,11 g/mol Bod varu: 433 °C (1013 hPa) Bod tání: 70…73 °C |
MDL Number:
MFCD00005904 CAS číslo: 5460-29-7 EINECS: 226-738-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...