1-Phenyl-1,3-butandione, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1-Phenylbutane-1,3-dione, 1-Benzoylacetone, Benzoylacetone, 1-Phenyl-1,3-butanedione
M-Clarity™ quality level = MQ100
B11907-10G
B11907-250G
B11907-50G
SIALB11907-10GEA
1700
CZK
SIALB11907-10G
SIALB11907-250G
SIALB11907-50G
1-Phenyl-1,3-butandione, Sigma-Aldrich®
1-Phenyl-1,3-butandione
Vzorec:
C₆H₅C(O)CH₂C(O)CH₃ MW: 162,19 g/mol Bod varu: 262 °C (1013 hPa) Bod tání: 57…59 °C Hustota: 1,0899 g/cm³ (60 °C) |
MDL Number:
MFCD00008786 CAS číslo: 93-91-4 EINECS: 202-286-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...