5-Amino-1H-tetrazole monohydrate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
A80602-100G
A80602-500G
SIALA80602-100GEA
4110
CZK
SIALA80602-100G
SIALA80602-500G
5-Amino-1H-tetrazole monohydrate, Sigma-Aldrich®
5-Amino-1H-tetrazole monohydrate
Vzorec:
CH₃N₅·1H₂O Bod tání: 216…217 °C Hustota: 1,51 g/cm³ (20 °C) |
MDL Number:
MFCD00149327 CAS číslo: 15454-54-3 EINECS: 224-581-7 UN: 1325 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...