3-(Aminomethyl)pyridine, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
3-Pyridylmethylamine, (3-Pyridyl)methylamine, 3-Picolamine, Pyridin-3-ylmethanamine, 3-Picolylamine
M-Clarity™ quality level = MQ100
A65409-100G
A65409-5G
SIALA65409-100GEA
2260
CZK
SIALA65409-100G
SIALA65409-5G
3-(Aminomethyl)pyridine, Sigma-Aldrich®
3-(Aminomethyl)pyridine
Vzorec:
C₆H₈N₂ MW: 108,14 g/mol Bod varu: 226 °C (1013 hPa) Bod tání: –21 °C Hustota: 1,0712 g/cm³ (20 °C) |
MDL Number:
MFCD00006412 CAS číslo: 3731-52-0 EINECS: 223-091-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...