2-Amino-4,6-dichloropyrimidine, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
A48601-100G
A48601-10G
SIALA48601-100GEA
28770
CZK
SIALA48601-100G
SIALA48601-10G
2-Amino-4,6-dichloropyrimidine, Sigma-Aldrich®
2-Amino-4,6-dichloropyrimidine
Vzorec:
C₄H₃Cl₂N₃ MW: 163,99 g/mol Bod tání: 216…220 °C |
MDL Number:
MFCD00006090 CAS číslo: 56-05-3 EINECS: 200-253-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...