Tisk…

1-Hexyl-3-methylimidazolium hexafluorophosphate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1-Hexyl-3-methyl-1H-imidazol-3-ium hexafluorophosphate(V)

M-Clarity™ quality level = MQ100

89320-50G 89320-5G
SIAL89320-50GEA 19670 CZK
SIAL89320-50G SIAL89320-5G
1-Hexyl-3-methylimidazolium hexafluorophosphate, Sigma-Aldrich®
1-Hexyl-3-methylimidazolium hexafluorophosphate
Vzorec: C₁₀H₁₉F₆N₂P
MW: 312,24 g/mol
Hustota: 1,28 g/cm³ (20 °C)
MDL Number: MFCD03093296
CAS číslo: 304680-35-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR