Tisk…

2-Bromo-p-toluonitrile, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

697206-25G 697206-5G
SIAL697206-25GEA 2660 CZK
SIAL697206-25G SIAL697206-5G
2-Bromo-p-toluonitrile, Sigma-Aldrich®
2-Bromo-p-toluonitrile
Vzorec: C₈H₆BrN
MW: 196,05 g/mol
MDL Number: MFCD00158968
CAS číslo: 42872-73-1
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR