Butyl cyanoacetate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Cyanoacetic acid butyl ester
M-Clarity™ quality level = MQ100
691283-1KG
691283-200KG
SIAL691283-1KGEA
0
CZK
SIAL691283-1KG
SIAL691283-200KG
Butyl cyanoacetate, Sigma-Aldrich®
Butyl cyanoacetate
Vzorec:
C₇H₁₁NO₂ MW: 141,17 g/mol Bod varu: 230,7 °C (1013 hPa) Bod tání: –80 °C Hustota: 1,001 g/cm³ (20 °C) |
MDL Number:
MFCD00001941 CAS číslo: 5459-58-5 EINECS: 226-730-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...