Tisk…

Ethyl benzoylacetate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Ethyl 3-oxo-3-phenylpropanoate

M-Clarity™ quality level = MQ100

690082-1KG 690082-200KG 690082-25KG
SIAL690082-1KGEA 7420 CZK
SIAL690082-1KG SIAL690082-200KG SIAL690082-25KG
Ethyl benzoylacetate, Sigma-Aldrich®
Ethyl benzoylacetate
Vzorec: C₁₁H₁₂O₃
MW: 192,21 g/mol
Bod varu: 265 °C (1013 hPa)
Bod tání: < 0 °C
Hustota: 1,114 g/cm³ (20 °C)
Bod vzplanutí: 140 °C
MDL Number: MFCD00009196
CAS číslo: 94-02-0
EINECS: 202-295-3
Merck Index: 13,03803

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR