6-Bromo-m-toluidine, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
686409-25G
686409-5G
SIAL686409-25GEA
11930
CZK
SIAL686409-25G
SIAL686409-5G
6-Bromo-m-toluidine, Sigma-Aldrich®
6-Bromo-m-toluidine
Vzorec:
C₇H₈BrN MW: 186,05 g/mol |
MDL Number:
MFCD01806278 CAS číslo: 53078-85-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...