Tisk…

4-Amino-2,6-dichloropyrimidine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2,6-Dichloropyrimidin-4-ylamine

M-Clarity™ quality level = MQ100

679070-25G 679070-5G
SIAL679070-25GEA 6330 CZK
SIAL679070-25G SIAL679070-5G
4-Amino-2,6-dichloropyrimidine, Sigma-Aldrich®
4-Amino-2,6-dichloropyrimidine
Vzorec: C₄H₃Cl₂N₃
MW: 163,99 g/mol
MDL Number: MFCD00038015
CAS číslo: 10132-07-7
EINECS: 233-369-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR