2,6-Bis(trifluoromethyl)pyridine, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
665649-1G
665649-250MG
SIAL665649-1GEA
0
CZK
SIAL665649-1G
SIAL665649-250MG
2,6-Bis(trifluoromethyl)pyridine, Sigma-Aldrich®
2,6-Bis(trifluoromethyl)pyridine
Vzorec:
C₇H₃F₆N MW: 215,1 g/mol |
MDL Number:
MFCD00236675 CAS číslo: 455-00-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...