3,5-Bis(trifluoromethyl)pyridine, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
665630-1G
665630-250MG
SIAL665630-1GEA
12870
CZK
SIAL665630-1G
SIAL665630-250MG
3,5-Bis(trifluoromethyl)pyridine, Sigma-Aldrich®
3,5-Bis(trifluoromethyl)pyridine
Vzorec:
C₇H₃F₆N MW: 215,1 g/mol Bod varu: 117…118 °C (1013 hPa) |
MDL Number:
MFCD00128904 CAS číslo: 20857-47-0 UN: 2810 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...