2-Hexynoic acid, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
662895-25G
662895-5G
SIAL662895-25GEA
0
CZK
SIAL662895-25G
SIAL662895-5G
2-Hexynoic acid, Sigma-Aldrich®
2-Hexynoic acid
Vzorec:
C₆H₈O₂ MW: 112,13 g/mol Bod varu: 110…112 °C (10 mmHg) Bod tání: 23…26 °C |
MDL Number:
MFCD00014371 CAS číslo: 764-33-0 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...