1-(3-Fluorophenyl)-2-thiourea, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
N-(3-Fluorophenyl)thiourea, 3-Fluorophenylthiourea
M-Clarity™ quality level = MQ100
654825-1G
654825-5G
SIAL654825-1GEA
1650
CZK
SIAL654825-1G
SIAL654825-5G
1-(3-Fluorophenyl)-2-thiourea, Sigma-Aldrich®
1-(3-Fluorophenyl)-2-thiourea
Vzorec:
(C₆H₄F)NC(S)NH₂ MW: 170,21 g/mol Bod varu: 276 °C (1013 hPa) Bod tání: 116 °C |
MDL Number:
MFCD00041167 CAS číslo: 458-05-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...